Learn More
StatLab MasterTech Fouchet's Reagent

Description
- Used to stain tissue sections to visualize bile pigments, aiding in the study of liver and biliary tract tissues
- The reagent works by oxidizing bilirubin to biliverdin, resulting in a green color change when bilirubin is present
- This method is particularly useful for diagnosing liver diseases and for histological staining of tissue samples
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| Color | Colorless |
| Quantity | 1 pt. |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.