Learn More
StatLab MasterTech G. M. S. Special Stain Kit
SDP
Catalog No. 21010337
Change view
Click to view available options
Product Type:
GMS Stain Kit
Catalog No. Product Type
21-010-337 GMS Stain Kit
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-337 Supplier StatLab Supplier No. KTGMSLT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Stains for fungi and Pneumocystis jirovecii in just 15 min.
  • This kit includes a 15 min. microwave procedure and the traditional one hour and thirty minute procedure
  • The MasterTech G.M.S. Staining Kit is suitable in lieu of and comparable to the Grocott Methenamine Silver Technique and differs from this technique through the use of a stabilized, pre-measured Methenamine Silver Compound, instead of Methenamine Borate, Hexamine, or Silver Nitrate Solutions; which benefits the end user through ease of use and a longer shelf life
  • A Methenamine Silver Developer Solution serves as the primary stain, with Gold Chloride toning the tissue sections, producing gray to black fungi and Pneumocystis jirovecii against a green background counterstained with Light Green
  • Liter Kit Consists of: Methenamine Silver Compound 40 vials (10 vials make 500 mL working solution, includes Sodium Borate), 5% Chromic Acid liter, 1% Sodium Bisulfite liter, 0.3% Gold Chloride liter, 2% Sodium Thiosulfate liter, Light Green Counterstain liter and plastic slide jars
  • Liter Stain Kit yields between 240-800 slides; slide yields assume maximum slides stained per run, maximum runs per use - Actual Slide Yields may vary
  • A MasterTech Special Stain Kit
Specifications
Volume (Metric) 1 L
Product Type GMS Stain Kit