Learn More
StatLab MasterTech Gentian Violet

Description
- Gentian violet, also known as crystal violet, is a triarylmethane dye used in histology for staining and classifying Gram-positive bacteria
- It is particularly useful in Gram's stain, where it helps in the metachromatic demonstration of amyloid and the primary classification of bacteria
- The dye's darker blue shade makes it the preferred choice for this purpose
- Gentian violet has antibacterial, antifungal, and anthelmintic properties, and was previously important as a topical antiseptic
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| Color | Violet |
| Quantity | 1 pt. |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.