Learn More
StatLab MasterTech Giemsa Stain, Stock
SDP
Catalog No. 22023751
Change view
Click to view available options
Quantity:
1 pt.
Catalog No. Quantity
22-023-751 1 pt.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-023-751 Supplier StatLab Supplier No. STGIEPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech Giemsa Stain, Stock - For detecting and diagnosing malaria and other blood parasites
  • Giemsa stain is the most commonly used stain for staining blood films for malaria diagnosis
  • It is used to differentiate the nuclear and cytoplasmic morphology of various blood cells, including red blood cells, white blood cells and platelets
  • It can differentiate human cells as purple and bacterial cells as pink
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 1 pt.
Product Type Giemsa Stain