Learn More
StatLab MasterTech Gill's 1 Hematoxylin
Description
- The MasterTech Line features traditional hematoxylins that are optimized for performance
- This single strength formulation optimally stains Gynecological and Non-Gynecological specimens
- Gill 1 is a general purpose progressive nuclear stain that is also useful as a nuclear counterstain for Special stains and IHC
Specifications
Specifications
| Type | Hematoxylin 1 |
| Product Type | Hematoxylin |
| Quantity | 1 gal. |