Learn More
StatLab MasterTech Gill's 1 Hematoxylin
SDP
Catalog No. 22033321
Change view
Click to view available options
Quantity:
1 gal.
5 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-033-321 5 gal.
21-033-377 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-033-321 Supplier StatLab Supplier No. HXGHE15CB
Add to Cart
Add to Cart
This item is not returnable. View return policy
The lightest of three Gill's Hematoxylin formulations and is best suited for staining in cytology preparations
  • The MasterTech Line features traditional hematoxylins that are optimized for performance
  • This single strength formulation optimally stains Gynecological and Non-Gynecological specimens
  • Gill 1 is a general purpose progressive nuclear stain that is also useful as a nuclear counterstain for Special stains and IHC
Specifications
Quantity 5 gal.
Product Type Hematoxylin 1