Learn More
StatLab MasterTech Gill's 2 Hematoxylin
Description
- The MasterTech Line features traditional hematoxylins that are optimized for performance.
- The higher dye concentration of Gill 2 is better suited for routine HandE staining, producing a darker nucleus that allows for shorter staining times than Gill 1
- This progressive stain produces distinct nuclear detail with great clarity
- Use for automated or hand staining as well as frozen sections.
Specifications
Specifications
| Product Type | Gill Hematoxylin (2X) |