Learn More
StatLab MasterTech Gill's 3 Hematoxylin
SDP
Catalog No. 22033322
Change view
Click to view available options
Product Type:
Hematoxylin 3
Catalog No. Product Type
22-033-322 Hematoxylin 3
1 options
This item is not returnable. View return policy
Catalog No. 22-033-322 Supplier StatLab Supplier No. HXGHE3LT
Add to Cart
Add to Cart
This item is not returnable. View return policy
With the highest dye concentration Gill 3 is ideally suited for routine H and E staining
  • The MasterTech Line features traditional hematoxylins that are optimized for performance
  • With the highest dye concentration Gill 3 is ideally suited for routine HandE staining, producing a darker nucleus with shorter staining times than either Gill 1 or 2
  • This progressive stain produces distinct nuclear detail with great clarity
  • Use for automated or hand staining, as well as frozen sections
Specifications
Product Type Hematoxylin 3