Learn More
StatLab MasterTech Gill's Hematoxylin 1
SDP
Catalog No. 22021416
Change view
Click to view available options
Quantity:
1 L
Catalog No. Quantity
22-021-416 1 L
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-416 Supplier StatLab Supplier No. HXGHE1LT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech Gill's Hematoxylin 1 - the lightest of three Gills Hematoxylin formulations and is best suited for staining in cytology preparations
  • A specific formulation of hematoxylin stain commonly used as a nuclear stain in routine H and E staining, particularly in cytology applications due to its lower dye concentration
  • Allows for delicate and precise staining of cell nuclei especially in gynecological and non-gynecological smears
  • It is considered a single strength hematoxylin ideal for cytological analysis
Specifications
Quantity 1 L
Product Type Gill 1 Hematoxylin