Learn More
StatLab MasterTech Gill's Hematoxylin 2
SDP
Catalog No. 22021380
Change view
Click to view available options
Quantity:
1 L
Catalog No. Quantity
22-021-380 1 L
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-380 Supplier StatLab Supplier No. HXGHE2LT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech Gill's Hematoxylin 2 - formulated for staining in most histology preparations
  • A specific type of hematoxylin stain primarily used for nuclear staining in both histology and cytology applications often preferred when a darker more intense nuclear stain is needed
  • Particularly effective for counterstaining in immunohistochemistry or when examining cytology samples
  • It can be used as a progressive or regressive stain depending on the staining time
Specifications
Quantity 1 L
Product Type Gill 2 Hematoxylin