Learn More
StatLab MasterTech Gill's Hematoxylin 3
SDP
Catalog No. 22000041
Change view
Click to view available options
1 gal.
Catalog No.
22-000-041 1 gal.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-041 Supplier StatLab Supplier No. HXGHE3GAL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
The MasterTech Line features traditional hematoxylins that are optimized for performance
  • Ideally suited for routine H and E staining, producing a darker nucleus with shorter staining times than either Gill 1 or 2
  • This progressive stain produces distinct nuclear detail with great clarity
  • Use for automated or hand staining, as well as frozen sections
Specifications
Product Type Gill's Hematoxylin 3