Learn More
StatLab™ MasterTech GMS Stain Kit
SDP
Catalog No. STLKTGMS
Change view
Click to view available options
Quantity:
100 mL
1 pt.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLKTGMS 100 mL
STLKTGMSPT 1 pt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLKTGMS Supplier StatLab™ Supplier No. KTGMS
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
GMS Stain Kit for fungi and Pneumocystis jirovecii stains in just 15 minutes.
  • Kit includes a 15 minute microwave procedure and the traditional one hour and thirty minute procedure
  • A Methenamine Silver Developer Solution serves as the primary stain, with Gold Chloride toning the tissue sections, producing gray to black fungi and Pneumocystis jirovecii against a green background counterstained with Light Green
  • 100 mL Stain Kit yields between 20 and 32 slides
  • 1 pt. Stain Kit yields between 100 and 400 slides
Specifications
Quantity 100 mL
Product Type Stain Kit