Learn More
StatLab MasterTech GMS Stain Kit
SDP
Catalog No. 22021432
Change view
Click to view available options
Quantity:
1 pt.
Catalog No. Quantity
22-021-432 1 pt.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-432 Supplier StatLab Supplier No. KTGMGPT
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech GMS Stain Kit - stains for bone marrow, core sections, mast cells and Helicobacter pylori in 25 minutes
  • This procedure is used to differentiate cells present in hematopoietic tissue
  • Both May-Grunwald and Giemsa Stock serve as primary stains producing blue to purple nuclei, cytoplasm, Helicobacter pylori, and mast cells
  • Stain Kit yields 54 slides and actual slide yields may vary
  • Pint Kit Consists of Giemsa Stain Stock Pint, May-Grunwald Stain Stock Pint. and 6.8 pH Phosphate Buffer Pint
  • All kit components should be stored in a dry environment between 15°C and 30°C
  • Please review all component labels to ensure correct storage temperature
  • A MasterTech Special Stain Kit
Specifications
Quantity 1 pt.
Product Type Modified Bielschowsky's Stain