Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Hall's Bile Stain Kit
SDP

Catalog No. 22021400
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-400 100 mL
22-021-381 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-400 Supplier StatLab Supplier No. KTHBI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Harmful if swallowed, in contact with skin, or if inhaled

StatLab MasterTech Hall's Bile Stain Kit - Stains for bile or bilirubin in less than 15 minutes

  • Oxidization of the bile with Fouchets Reagent provides the primary stain producing green bile or bilirubin with pink to red collagen and a tan to yellow background of muscle and cell cytoplasm and counterstained by Van Gieson's
  • 100 mL Stain Kit yields between 80 and 100 slides and consists of - Fouchets Reagent 100 mL and Van Giesons Stain 100 mL
  • All kit components are available for purchase individually and the kit is sealed with a tamper-evident label for quality assurance
  • All kit components should be stored in a dry environment between 15°C and 30°C
  • Please review all component labels to ensure correct storage temperature
  • StatLab recommends using a Bile Control Slide
  • A MasterTech Special Stain Kit

Specifications

Quantity 100 mL
Product Type Hall's Bile Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.