Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Harris Hematoxylin
SDP

Catalog No. 22033324
Change view
Click to view available options
Quantity:
16 oz.
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-033-324 16 oz.
22-033-323 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-033-324 Supplier StatLab Supplier No. HXHHEPT
Add to Cart
Add to Cart
This item is not returnable. View return policy

One of the original hematoxylin formulations, provides deep rich nuclear staining

  • Typically used as a regressive stain with MasterTech differentiating solution, this mercury free formulation is ideal for Histology applications
  • Used in conjunction with Eosin for HandE staining to screen specimens in anatomic pathology, research, smears, touch preps and other applications

Specifications

For Use With (Equipment) Staining Equipment
Grade Chemical Solution
Product Type Hematoxylin
Quantity 16 oz.
Solution Type Stain
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.