Learn More
StatLab MasterTech Harris Hematoxylin
SDP
Catalog No. 22021436
Change view
Click to view available options
Quantity:
1 gal.
Catalog No. Quantity
22-021-436 1 gal.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-436 Supplier StatLab Supplier No. HXHHEGAL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech Harris Hematoxylin - Formulated to give superior detail every time
  • Used as a nuclear stain specifically to stain the nuclei of cells within tissue samples providing clear detail of the nuclear structure and allowing for better visualization of cell morphology during microscopic examination
Specifications
Quantity 1 gal.
Product Type Harris Hematoxylin