Learn More
StatLab MasterTech Hematoxylin Powder
SDP
Catalog No. 21010256
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
21-010-256 100 g
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-256 Supplier StatLab Supplier No. AHHEM100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
A natural dye derived from the heartwood of the logwood tree and is the most commonly used stain in histology.
  • Hematoxylin powder can be formulated into an aqueous stain and used to permanently stain tissues
  • Hematoxylin powder is oxidized to hematein, which binds to nucleic acids, especially the nuclear chromatin of cells, providing a deep blue to purple coloration
  • 100 g powder
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 100 g
Product Type Hematoxylin