Learn More
StatLab MasterTech Herovici's Collagen Stain Kit
SDP
Catalog No. 22021379
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-379 100 mL
22-021-378 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-379 Supplier StatLab Supplier No. KTHER
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech Herovicis Collagen Stain Kit - Stains for young and mature collagen in less than 10 minutes
  • Used to differentiate young and mature collagen as well as stain reticulum
  • Herovici's working solution stains young collagen and reticulum blue and mature collagen red while providing a yellow cytoplasm counterstain
  • Nuclei are stained blue to black with Weigerts Hematoxylin
  • 100 mL Stain Kit yields between 36 and 45 slides and all kit components should be stored in a dry environment between 15°C and 30°C
  • StatLab recommends using a Skin (Normal) Control Slides
  • A MasterTech Special Stain Kit
Specifications
Quantity 100 mL
Product Type Herovici's Collagen Stain