Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Hyaluronidase Powder
SDP

Catalog No. 21033267
Change view
Click to view available options
Color:
Light Brown
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-033-267 Light Brown
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-267 Supplier StatLab Supplier No. AHHYA100
Add to Cart
Add to Cart

Often used in conjunction with collagenase to dissociate the extracellular matrix between cells of animal tissue

  • It is often used in conjunction with collagenase to dissociate the extracellular matrix between cells of animal tissue, in order to release viable cells for use in tissue culture
  • It may also be used to clarify synovial fluids in order to make cell counts possible
  • Type I-S, lyophilized powder, 400-1000 units/mg solid
  • Activity: 300 i.u./mg
  • A MasterTech Special Stain Kit Component

Specifications

Physical Form Powder
CAS 37326-33-3
Color Light Brown
Quantity 100 mg
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.