Learn More
StatLab MasterTech India Ink
SDP
Catalog No. 21033465
Change view
Click to view available options
Volume (Metric) Exhaust:
25 mL
Catalog No. Volume (Metric) Exhaust
21-033-465 25 mL
1 options
Catalog No. 21-033-465 Supplier StatLab Supplier No. STIIN25
Add to Cart
Add to Cart
For marking pathology specimens, the technique involves gently painting the ink onto the tissue with a fine brush or applicator
  • Ink is typically applied directly to the surface of fresh tissue specimens
  • The technique involves gently painting the ink onto the tissue with a fine brush or applicator
  • Once applied, the ink is allowed to dry before the tissue is processed for further histological examination
  • The carbon particles in the ink adhere to the tissue surface, providing a persistent mark that remains visible throughout the fixation, embedding, and sectioning processes
  • A MasterTech Tissue Marking Dye
Specifications
Volume (Metric) Exhaust 25 mL