Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Iron Special Stain Kit
SDP

Catalog No. 21010339
Change view
Click to view available options
Quantity:
1 L
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-010-339 1 L
21-010-338 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-339 Supplier StatLab Supplier No. KTIROLT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Stains ferric iron in under 30 min.

  • This kit is suitable in lieu of and comparable to the Perl’s Technique and Gomori’s Iron Reaction for staining ferric iron as the MasterTech Iron Stain is less hazardous than these procedures and provides more precise results by using lower strength Hydrochloric Acid and Potassium Ferrocyanide
  • A Working Prussian Blue Solution serves as the primary stain, producing blue ferric iron against a pink to red background with red nuclei, counterstained by Nuclear Fast Red
  • There is no filtration required when using MasterTech Nuclear Fast Red
  • Gallon Stain Kit yields between 2800-3800 slides; Slide Yields assume maximum slides stained per run, maximum runs per use - Actual Slide Yields may vary
  • Gallon Kit Consists of: 3% Potassium Ferrocyanide 2 gallons, 3% Hydrochloric Acid 2 gallons, and Nuclear Fast Red Stain gallons
  • A MasterTech Special Stain Kit

Specifications

Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.