Learn More
StatLab™ MasterTech Iron Stain Kit
Description
- A Working Prussian Blue Solution serves as the primary stain, producing blue ferric iron against a pink to red background with red nuclei, counterstained by Nuclear Fast Red
- No filtration required when using MasterTech Nuclear Fast Red
- 100 mL Stain Kit yields between 80 and 100 slides
- 1 pt. Stain Kit yields between 320 and 500 slides
Specifications
Specifications
| Quantity | 100 mL |
| Product Type | Stain Kit |