Learn More
StatLab MasterTech Luxol Fast Blue-Cresyl Echt Violet Stain Kit
SDP
Catalog No. 21033399
Change view
Click to view available options
Quantity:
100 mL
16 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-033-399 16 oz.
21-033-398 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-033-399 Supplier StatLab Supplier No. KTLFBPT
Add to Cart
Add to Cart
Stains myelin sheaths and nissl substances in one hour and fifteen minutes
  • MasterTechs improved Luxol Fast Blue Stain Kit contains a specially formulated Luxol Fast Blue stain that reduces staining time to one hour and fifteen minutes, while most other kits on the market take over two hours!
  • Both Luxol Fast Blue and Cresyl Echt Violet serve as primary stains, producing vibrant blue myelin fibers, with dark blue nissl substances and blue nuclei
  • 100 mL Stain Kit yields between 53 and 66 slides
  • 100 mL Kit Consists of: Luxol Fast Blue Stain 100 mL, Cresyl Echt Violet Stain 100 mL, 70% Reagent Alcohol Pint, 0.05% Lithium Carbonate Pint
  • A MasterTech Special Stain Kit
Specifications
Quantity 16 oz.
Product Type Luxol Fast Blue Standard Histology Control Slide