Learn More
StatLab MasterTech Luxol Fast Blue Stain

Description
- It is effective in differentiating between myelinated and unmyelinated nerve fibers, which is crucial for understanding the structure and function of the nervous system
- The stain works by binding to the negatively charged components of myelin sheaths, which are rich in lipoproteins
- This binding allows for the visualization of myelinated regions, which appear blue under the stain
- It can also be used to detect demyelination and remyelination processes, providing insights into various neurological conditions
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| Chemical Name or Material | Luxol Fast Blue Stain |
| Grade | Histological |
| Color | Blue |
| Quantity | 100 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.