Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Luxol Fast Blue Stain
SDP

Catalog No. 22021444
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-444 500 mL
22-021-373 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-444 Supplier StatLab Supplier No. STLFBPT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab MasterTech Luxol Fast Blue Stain - for identifying and visualizing myelin in the central nervous system CNS

  • It is effective in differentiating between myelinated and unmyelinated nerve fibers, which is crucial for understanding the structure and function of the nervous system
  • The stain works by binding to the negatively charged components of myelin sheaths, which are rich in lipoproteins
  • This binding allows for the visualization of myelinated regions, which appear blue under the stain
  • It can also be used to detect demyelination and remyelination processes, providing insights into various neurological conditions
  • A MasterTech Special Stain Kit Component

Specifications

Chemical Name or Material Luxol Fast Blue Stain
Grade Histological
Color Blue
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.