Learn More
StatLab MasterTech McDonald's Gram Stain Kit
SDP
Catalog No. 21033402
Change view
Click to view available options
Volume (Metric):
16 oz.
Catalog No. Volume (Metric)
21-033-402 16 oz.
1 options
Catalog No. 21-033-402 Supplier StatLab Supplier No. KTMGSPT
Add to Cart
Add to Cart
Stains for gram positive and gram negative bacteria in less than five minutes
  • This kit is suitable in lieu of and comparable to the Brown and Brenn, Brown and Hopps, and Modified Gram Methods for staining gram positive and gram negative bacteria
  • The MasterTech Gram Stain Kit differs from these procedures in that Carbol Fuchsin is used instead of Saffranin which intensifies the stains
  • The kit is also safer to use than the traditional procedures because the hazardous Gallegos Differentiating Solution and picric acid/acetone is eliminated
  • The results are identical to the Brown and Brenn stain without the exposure to hazardous chemicals
  • Both Gentian Violet and Carbol Fuchsin serve as the primary stains, producing blue gram positive bacteria and red gram negative bacteria against a yellow background, counterstained by Optimized Tartrazine which provides more intense staining in less time than traditional Tartrazine formulations
  • Pint Stain Kit yields between 144-225 slides
  • Pint Kit Consists of: Gentian Violet Pint, Universal Iodine Solution Pint, Gram's Decolorizer Pint, Carbol Fuchsin Pint and Optimized Tartrazine Counter Stain Pint
  • A MasterTech Special Stain Kit
Specifications
Volume (Metric) 16 oz.
Product Type McDonald's Gram