Learn More
StatLab MasterTech Melanin Bleach Kit
Description
- Melanin bleaching is commonly used to bleach Melanin in tissue so that other cells that look like Melanin can be distinguished and observed when stained
- This kit is fast and effective in removing melanin from hair, skin, retina or substantia nigra of the brain
- In comparison to other bleach kits, the MasterTech Melanin Bleach Kit is safer to use because it does not contain hazardous sulfuric acid
- 100 mL Stain Kit yields 80-100 slides
- Kit Consists of: 1% Potassium Permanganate 100 mL and 2% Oxalic Acid 100 mL
- A MasterTech Special Stain Kit
Specifications
Specifications
| Form | Liquid |
| Product Type | Special Stain Kit |