Learn More
StatLab MasterTech Metanil Yellow Stain
SDP
Catalog No. 21060044
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
21-060-044 100 mL
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-060-044 Supplier StatLab Supplier No. STMYE100
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
Used to stain collagen yellow in contrast to the red muscle.
  • The stain is intended to identify epithelial mucins by staining background tissue shades of yellow when used in a mucicarmine stain procedure
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 100 mL
Product Type Alizarin Red