Learn More
StatLab MasterTech Methenamine Granular
SDP
Catalog No. 22021395
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
22-021-395 500 g
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-395 Supplier StatLab Supplier No. AHMET500
Harmful if swallowed, in contact with skin, or if inhaled
StatLab Methenamine Granular
  • Used as a component of the Grocotts methenamine silver stain (GMS) which is a special stain specifically designed to identify and visualize fungal organisms within tissue samples particularly highlighting the cell walls of fungi by staining them black against a lighter background making it a key tool for diagnosing fungal infections in pathology examinations
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 500 g
Product Type Methenamine/Borax