Learn More
StatLab MasterTech Methylene Blue Stain

Description
- A versatile and widely used dye in histology for various purposes
- The dye's ability to bind to negatively charged components, such as DNA and RNA, allows for detailed examination of cellular morphology and organization
- It is also used as a counterstain in Gram staining procedures, aiding in the differentiation of bacterial types
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| Chemical Name or Material | Methelyne Blue Counterstain |
| CAS | 7220-79-3 |
| Color | Blue |
| Quantity | 100 mL |
| Grade | Histological |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.