Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Methylene Blue Stain
SDP

Catalog No. 22021408
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-408 500 mL
22-021-365 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-408 Supplier StatLab Supplier No. STMBLPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab MasterTech Methylene Blue Stain - effective in staining nucleic acids, which are crucial for visualizing cell structures and differentiating between live and dead cells

  • A versatile and widely used dye in histology for various purposes
  • The dye's ability to bind to negatively charged components, such as DNA and RNA, allows for detailed examination of cellular morphology and organization
  • It is also used as a counterstain in Gram staining procedures, aiding in the differentiation of bacterial types
  • A MasterTech Special Stain Kit Component

Specifications

Chemical Name or Material Methelyne Blue Counterstain
Grade Histological
Color Blue
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.