Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Modified Bielschowsky's Stain Kit
SDP

Catalog No. 22021443
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-443 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-443 Supplier StatLab Supplier No. KTBIE
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

StatLab MasterTech Modified Bielschowsky's Stain Kit - stains axons, intracellular neurofibrils, and senile plaques in just 35 minutes

  • This kit is suitable in lieu of and comparable to the Gros-Bielschowskis, Sevier-Munger, Rio-Hortega, and Holmes procedures for staining axons
  • Silver Nitrate serves as both the primary stain and counterstain producing black axons, neurofibrils, and senile plaques against a tan to gold background
  • 100 mL Stain Kit yields between 30 and 96 slides and actual yield slides may vary
  • 100 mL Kit Consists of - Silver Nitrate Solution Pint, 5% Sodium Thiosulfate 100 mL, Stock Developer Solution A 6 mL, Stock Developer Solution B 6 mL, Stock Developer Solution C 6 mL
  • One or more component(s) in this kit require a storage temperature between 2°C and 10°C
  • A MasterTech Special Stain Kit

Specifications

Quantity 100 mL
Product Type Modified Bielschowsky's Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.