Learn More
StatLab MasterTech Modified Brown-Hopps Special Stain Kit
SDP
Catalog No. 21010336
Change view
Click to view available options
Product Type:
Gram's Stain Kit
Catalog No. Product Type
21-010-336 Gram's Stain Kit
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-336 Supplier StatLab Supplier No. KTBHGPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Stains for gram positive and gram negative bacteria in just under 20 min.
  • This kit is suitable in lieu of and comparable to various Brown-Hopps staining procedures
  • Basic Fuchsin and Crystal Violet both serve as primary stains, producing blue stained gram positive bacteria, along with red gram negative bacteria and nuclei, while the Picric Acid/Acetone provides a yellow counterstain
  • Pint Kit Consists of: 1% Crystal Violet Pint, Universal Iodine Solution Pint, 0.1% Basic Fuchsin Pint, Gallego's Solution Pint and Picric Acid/Acetone Pint
  • Pint Stain Kit yields between 144-225 slides and slide yields assume maximum slides stained per run, maximum runs per use - actual slide yields may vary
  • A MasterTech Special Stain Kit
Specifications
Volume (Metric) 473 mL
Product Type Gram's Stain Kit