Learn More
StatLab MasterTech Modified Mayer's Hematoxylin
SDP
Catalog No. 22021403
Change view
Click to view available options
Quantity:
1 L
Catalog No. Quantity
22-021-403 1 L
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-403 Supplier StatLab Supplier No. HXMMHLT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech Modified Mayer's Hematoxylin - Requires no differentiation or separate bluing solution
  • Used in histology as a nuclear stain specifically as a counterstain in procedures like immunohistochemistry (IHC) to clearly visualize cell nuclei
Specifications
Quantity 1 L
Product Type Modified Mayer's Hematoxylin