Learn More
StatLab MasterTech Modified Mayers Hematoxylin
SDP
Catalog No. 21010297
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
21-010-297 100 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-297 Supplier StatLab Supplier No. HXMMH100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Requires no differentiation or separate bluing solution.
  • This modified version of Mayer's formulation offers distinct and striking nuclear detail
  • A light delicate hematoxylin with no mucin affinity, it requires no differentiating and no bluing step to achieve optimal results
  • Ready-to-use, no filtering is required
Specifications
Quantity 100 mL
Product Type Modified Mayer's Hematoxylin