Learn More
StatLab MasterTech Morse's Decalcifier
SDP
Catalog No. 21010285
Change view
Click to view available options
Form:
Solution
Catalog No. Form
21-010-285 Solution
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 21-010-285 Supplier StatLab Supplier No. DCM0150
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Provides rapid decalcification of tissue.
  • Provides rapid decalcification of tissue to maximize retention of RNA integrity for in situ hybridization (ISH) procedures while preserving morphology
  • Solution is particularly effective for ISH procedures
  • Morse's Solution is recommended for decalcifying tissues to be processed for the rapid ISH analysis of specific RNA expression
Specifications
Product Type Decalcifying Solution
Quantity 1 L
Solution Type Decalcifier
Form Solution