Learn More
StatLab MasterTech Muci-10-Plus
SDP
Catalog No. 22024801
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
22-024-801 100 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-024-801 Supplier StatLab Supplier No. STMPL100
Add to Cart
Add to Cart
This item is not returnable. View return policy
A Rapid Staining Solution to detect Epithelial Mucins and highlight epithelial mucins in tissue sections.
  • It acts as a specialized, fast-acting alternative to the traditional Mayer's Mucicarmine method, reducing staining time to 10 minutes.
  • A StatLab MasterTech Special Stain Kit Component
Specifications
Quantity 100 mL