Learn More
StatLab MasterTech Muci-10 Red Stain Concentrate
SDP
Catalog No. 22000084
Change view
Click to view available options
25 mL
Catalog No.
22-000-084 25 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-000-084 Supplier StatLab Supplier No. STMCO25
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
10 drops in 20ml of water turns Mucin or Cryptococcus red in 10 minutes
  • 10 drops in 20 mL of water turns Mucin or Cryptococcus red in 10 minutes
  • A MasterTech Special Stain Kit Component
Specifications
Product Type Neutral Red