Learn More
StatLab™ MasterTech Muci Plus Stain Kit
SDP
Catalog No. STLKTMPLPT
Change view
Click to view available options
Quantity:
100 mL
1 pt.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLKTMPLPT 1 pt.
STLKTMPL 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLKTMPLPT Supplier StatLab™ Supplier No. KTMPLPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Muci Plus Stain Kit stains epithelial mucins in just 20 minutes.
  • Muci-10 Plus serves as the primary stain, producing pink to red epithelial mucins
  • Hematoxylin acts as a secondary stain, producing blue to purple nuclei against a yellow background, counterstained by Metanil Yellow
  • Kit is suitable in lieu of and comparable to the Mayer Mucicarmine method for staining mucins
  • 100 mL Stain Kit yields between 80 and 100 slides
  • 1 pt. Stain Kit yields between 320 and 500 slides
Specifications
Quantity 1 pt.
Product Type Stain Kit