Learn More
StatLab™ MasterTech Mucicarmine Stain
SDP
Catalog No. STLSTMUC100
Change view
Click to view available options
Quantity:
100 mL
1 pt.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
STLSTMUC100 100 mL
STLSTMUCPT 1 pt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSTMUC100 Supplier StatLab™ Supplier No. STMUC100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Mucicarmine Stain stains Mucin or Cryptococcus red in 1 to 2 hours.
  • Used in histology to identify mucin
Specifications
Quantity 100 mL
Product Type Mucicarmine Stock Solution