Learn More
StatLab™ MasterTech Mucicarmine Stain
SDP
Catalog No. STLSTMUCPT
Change view
Click to view available options
Quantity:
1 pt.
Catalog No. Quantity
STLSTMUCPT 1 pt.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLSTMUCPT Supplier StatLab™ Supplier No. STMUCPT
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Mucicarmine Stain stains Mucin or Cryptococcus red in 1 to 2 hours.
  • Used in histology to identify mucin
Specifications
Quantity 1 pt.
Product Type Mucicarmine Stock Solution