Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ MasterTech Nuclear Fast Red Stain
SDP

Catalog No. STLSTNFRLT
Change view
Click to view available options
Quantity:
1 L
1 gal./Bottle, 4 Bottles/Cs.
1 pt.
100 mL
5 gal. Bottle
Catalog No. Quantity
STLSTNFRLT 1 L
STLSTNFR100 100 mL
STLSTNFRPT 1 pt.
STLSTNFR5CB 5 gal. Bottle
STLSTNFRGAL 1 gal./Bottle, 4 Bottles/Cs.
5 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.

Catalog No. STLSTNFRLT

Supplier: StatLab™ STNFRLT

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.

Nuclear Fast Red Stain is a highly versatile reagent that selectively stains nuclear chromatin red, while the nonspecific signal is detected in pink tones.

  • For the staining of the nuclei where it will react with negatively charged components such as nucleic acids in the nucleus
  • An ideal alternative to hematoxylin which would otherwise obscure the substrate colors

Specifications

Type Nuclear Stain
Quantity 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.