Learn More
StatLab MasterTech Oil Red O Stain Kit

Description
- Oil Red O serves as the primary stain, producing red fat with blue nuclei stained by the Hematoxylin
- 100 mL Stain Kit yields between 80 and 100 slides
- 100 mL Kit Consists of: Propylene Glycol Pint, Oil Red Stain O 100 mL and Modified Mayers Hematoxylin 100 mL
- A MasterTech Special Stain Kit
Specifications
Specifications
| CAS | 64-19-7, 57-55-6 |
| Color | Red |
| Quantity | 16 oz. |
| Grade | Laboratory |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.