Learn More
StatLab MasterTech One Step Trichrome Stain (Blue and Red)
SDP
Catalog No. 22021425
Change view
Click to view available options
Quantity:
3.5 oz.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-425 3.5 oz.
22-021-364 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-425 Supplier StatLab Supplier No. STOSTB100
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech One Step Trichrome Stain (Blue and Red) - for differentiating between collagen and muscle fibers
  • This staining method allows pathologists and researchers to visualize tissue architecture with remarkable clarity, making it invaluable for identifying pathological conditions such as liver fibrosis, myocardial fibrosis, and tumor invasion patterns in cancer research
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 3.5 oz.
Product Type Trichrome One-Step Red