Learn More
StatLab MasterTech One-Step Trichrome Stain (Green and Red)
SDP
Catalog No. 22021419
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
22-021-419 100 mL
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-419 Supplier StatLab Supplier No. STOSTG100
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech One Step Trichrome Stain (Green and Red) - used for differentiating and visualizing various tissue components
  • It allows for the visualization of collagen fibers, muscle fibers, and cytoplasmic structures
  • Under the microscope, collagen fibers appear bright red, muscle fibers appear green, and cytoplasmic structures appear blue
  • This method is particularly useful in the diagnosis of diseases such as muscular dystrophy, cirrhosis, and fibrosis
  • It can also be used to assess the quality of collagen in tissues such as skin, bone, and cartilage
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 100 mL
Product Type Trichrome One-Step Green-Red