Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab MasterTech Optimized Giemsa Buffer
SDP

Catalog No. 21033275
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-033-275 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-275 Supplier StatLab Supplier No. BUG1043
Add to Cart
Add to Cart

Developed to increase metachromatic staining of bone marrow and other tissue sections

  • Specifically formulated to provide optimal staining for Bone Marrow and Gastric Tissue sections
  • It is used in place of standard Phosphate Buffer pH 6.8 in Giemsa, Giemsa/May Grunwald, and Wright-Giemsa procedures for these tissue types
  • A MasterTech Special Stain Kit Component

Specifications

For Use With (Equipment) Staining Equipment
Grade Chemical Solution
Product Type Giemsa Buffer
Quantity 16 oz.
Solution Type Buffer
For Use With (Application) Manual Special Staining
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.