Learn More
StatLab MasterTech Optimized Giemsa Buffer

Description
- Specifically formulated to provide optimal staining for Bone Marrow and Gastric Tissue sections
- It is used in place of standard Phosphate Buffer pH 6.8 in Giemsa, Giemsa/May Grunwald, and Wright-Giemsa procedures for these tissue types
- A MasterTech Special Stain Kit Component
Specifications
Specifications
| For Use With (Equipment) | Staining Equipment |
| Grade | Chemical Solution |
| Product Type | Giemsa Buffer |
| Quantity | 1 gal. |
| Solution Type | Buffer |
| For Use With (Application) | Manual Special Staining |
| pH | 6.8 |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.