Learn More
StatLab MasterTech Optimized Tartrazine Counterstain
SDP
Catalog No. 21033382
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
21-033-382 100 mL
1 options
Catalog No. 21-033-382 Supplier StatLab Supplier No. KC3251
Add to Cart
Add to Cart
Provides more intense staining in less time than traditional tartrazine formulations
  • Used as a yellow counterstain, most commonly in combination with a red dye like phloxine, to differentiate specific cellular components like viral inclusion bodies or Paneth cell granules within tissue sections, providing a contrasting color to the primary stain and highlighting the target structures with a yellow background color
  • A component in the MasterTech McDonalds Gram Stain Kit
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 100 mL
Product Type Tartrazine Solution