Learn More
StatLab MasterTech Optimized Tartrazine Counterstain
SDP
Catalog No. 22021390
Change view
Click to view available options
Quantity:
1 pt.
Catalog No. Quantity
22-021-390 1 pt.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-021-390 Supplier StatLab Supplier No. KC3252
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
StatLab MasterTech Optimized Tartrazine Counterstain - provides more intense staining in less time than traditional tartrazine formulations
  • Used as a yellow counterstain most commonly in combination with a red dye like phloxine to differentiate specific cellular components like viral inclusion bodies or Paneth cell granules within tissue sections providing a contrasting color to the primary stain and highlighting the target structures with a yellow background color
  • A component in the MasterTech McDonalds Gram Stain Kit
  • A MasterTech Special Stain Kit Component
Specifications
Quantity 1 pt.
Product Type Tartrazine Solution