Learn More
MasterTech Orcein Stain, StatLab
SDP
Catalog No. 22035375
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
22-035-375 100 mL
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-035-375 Supplier SLMP Llc Dba Statlab Medical Supplier No. KC1141
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled
A StatLab MasterTech Special Stain Kit Component.
  • A specialized histological dye primarily used to visualize elastic fibers (dark brown/purple), detect Hepatitis B surface antigen (HBsAg) in liver tissue (dark brown-purple), and identify copper-associated protein
  • It is a versatile stain for examining connective tissue architecture, skin lesions, and liver pathology
Specifications
Quantity 100 mL
Product Type Orcein Stain